Contec® PeridoxRTU®
Supplier: CONTEC
FBC0902PX FBC3702PX SBC0902PX SBC3702PX
CONTFBC0902PXEA 331 EUR
CONTFBC0902PX CONTFBC3702PX CONTSBC0902PX CONTSBC3702PX
Contec® PeridoxRTU®
Cleaning Agents Disinfectants
Blend of hydrogen peroxide and peracetic acid.
  • Patented hyperactive chemistry makes it fast acting so save time spent of biodecontamination
  • Validated sterile verison available makes it suitable for Grade A and B cleanrooms
  • Excellent material compatibility and will not damage common cleanroom materials
  • Double/single bagged helps facilitate transfer disinfection into cleanroom
  • Push/pull cap on 0.9L eliminates droplets reducing the risk of inhalation and can be easily applied to a wipe or cleaning tool
Contec PeridoxRTU is a BPR and EPA authorisedMore Product Information