Sample containers, snap-seal, Corning®
Supplier: Corning
1730-10 1730-2C 1730-4H 1730-4L 1730-8
CORN1730-10EA 87.29 EUR
CORN1730-10 CORN1730-2C CORN1730-4H CORN1730-4L CORN1730-8
Sample containers, snap-seal, Corning®
Sample Containers General Purpose Containers
PP, disposable, translucent.
  • Snap-Seal locking device keeps the cap closed and secure
  • Hinged cap reduces the chance of sample contamination
  • Graduations in both milliliters and ounces
  • Rough source for marking
These wide mouth sample containers are designed for a wide variety of applications and provide a reliable leak-tight seal when closed properly.
Upozornení: The product(s) on this page are outside the scope of the EU Directive 2019/904.