Clear bottom, black and white, polystyrene microplates, 384-well, Corning®
Supplier: Corning
3985 3643 3985BC 3540 3542 3544 3766 3762 3764 3768 3770 3764BC 3770BC 3765 3769
734-1200EA 1997.06 EUR
734-1200 734-4118 734-4168 734-4086 734-4087 734-4088 CORN3766 CORN3762 CORN3764 CORN3768 738-0210 CORN3764BC 738-0211 CORN3765 CORN3769
Clear bottom, black and white, polystyrene microplates, 384-well, Corning®
Microplates HTS Microplates
PS, flat bottom. Suitable for fluorescent and luminescent assays using either top or bottom detection plate readers.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification RECs, ISO 14001 - Kennebunk