Corning® Elplasia® 12K Open Well Plate
Supplier: Corning
4547
CORN4547EA 484.5 EUR
CORN4547
Corning® Elplasia® 12K Open Well Plate
Microplates Cell Culture Plates
Culture thousands of spheroids in a microplate footprint.
  • Ease of spheroid formation, culture, assessment, and harvest
  • Ready-to-use plate to create large quantities of uniform spheroids
  • Scaffold-free cultures
  • Culture spheroids for up to 14 or more days (cell line-dependent)
  • Highly reproducible bulk spheroid formation across microcavities
  • Common media reservoir for equivalent culture conditions for all spheroids
  • Easy access to spheroid cultures
  • Compatible with brightfield and fluorescent microscopy
Critical in drug screening and other applications suchMore Product Information