Steam-Thru II configuration couplings
Supplier: CPC COLDER
STC2021000
CPCCSTC2021000EA 1410 EUR
CPCCSTC2021000
Steam-Thru II configuration couplings
Fittings and Connectors
Steam-Thru II Connections offer the flexibility of 'steam on' and 'steam off' functionality. The innovative design allows the valve to be returned to the steam position enabling a second SIP cycle following media transfer.
Materials of construction include USP class 6 animal free (BSE/TSE) polysulphone, USP class 6 andimal free (BSE/TSE) silicone O-rings and seals.
Certifikace: USP Class VI, BSE/TSE free materials (ADCF).