E.Z.N.A.® Plasmid Midi kit

Supplier: OMEGA BIO-TEK

D6904-03 D6904-04 D6904-00
D6904-03EA 208 EUR
D6904-03 D6904-04 D6904-00
E.Z.N.A.® Plasmid Midi kit
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents

E.Z.N.A.® Plasmid Midi Kit offers a novel and reliable method of isolating up to 200 µg of plasmid DNA in less than 60 minutes. This kit combines HiBind® membrane technology with the time-tested consistency of alkaline SDS lysis of bacterial cells to deliver high-quality plasmid DNA. The DNase/RNase-free HiBind® DNA midi columns operate by eliminating time consuming phenol-chloroform extractions and alcohol precipitations. Plasmid purification using the E.Z.N.A.® Plasmid Midi Kit follows a simple bind-wash-elute procedure thereby making it possible for multiple samples to be processed. Although yields vary according to plasmid copy number, E.coli strain, and conditions of growth, 50 ml of an overnight culture in LB medium will typically produce 200 µg of high-copy number plasmid DNA. When working with low-copy number plasmids the starting culture volume can be increased up to 100 ml. High-quality DNA is ready for immediate use in routine molecular biology applications, such as automated fluorescent sequencing, restriction enzyme digestion, and transfection screening.


  • Purification of plasmid DNA in <60 minutes
  • No phenol/chloroform extractions
  • DNA suitable for most downstream applications

Order Now

SPECIFICATIONS

Description E.Z.N.A.® Plasmid Midi Kit I
Environmentally Preferable
Application
Target molecule Plasmid
Refine target molecule Plasmid
Sample type Bacteria
Method / Format Column (silica-based)
Prep size Midi
Sample size 15 - 50 ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR