Absorbing sheet for medical packaging
Supplier: DAKLAPACK
460583
DAKL460583EA 137 EUR
DAKL460583
Absorbing sheet for medical packaging
Mailers and Shipping Containers
These non-transparent absorbing materials are part of the packaging instruction P650. In emergencies the material will absorb all the liquid that has to stay inside the packaging. The absorbing sheets are on one side provided with a fluid-tight base. That ensures that the outside will stay clean and dry.