Face shields, Supervizor™
Supplier: HONEYWELL SAFETY
1002295 1002297 1002309 1002302 1002304 1002305 1002310 1002312 1002329 1002330 1004583 1004584 1004587
DALL1002295EA 13.5 EUR
DALL1002295 111-0727 DALL1002309 111-0549 DALL1002304 DALL1002305 DALL1002310 111-0728 DALL1002329 DALL1002330 DALL1004583 DALL1004584 DALL1004587
Face shields, Supervizor™
Face Shields
Robust, heavy-duty, helmet mounted face shield range with hearing protection compatibility. The products provide protection from abrasion, chemical hazards, gas, vapors and impact - they are therefore ideal for chemical applications or forestry.
  • Secure visor fixing methods allow easy visor replacement and maximum protection, allowing full unrestricted head movement
  • Items are compatible with a wide range of helmet designs and helmet mounted earmuffs (including Bilsom communication earmuffs)
  • Browguard with ratchet headband provides additional safety and