Eyewash bottle
Supplier: DELTALAB
19384
DELT19384EA 129 EUR
DELT19384
Eyewash bottle
First Aid Supplies Eyewash Stations and Eyewash Bottles
Made of low density polyethylene, this eyewash bottle is flexible and easy to squeeze.
  • Has an eye cup with snap-on cap
  • Eyecup has a perforated plate to break up the stream providing a gentle spray to the eye
  • Capped air vent valve prevents drawback of contaminated eye wash into the bottle
  • Contaminated solution exits through a drain tube, which rotates in any direction
  • Instructions for use printed in various languages