Intelsius® PharmaTherm Pallet Shippers
Supplier: DGP INTELSIUS
PHT375
DGINPHT375EA 212 EUR
DGINPHT375
Intelsius® PharmaTherm Pallet Shippers
Mailers and Shipping Containers
The Intelsius® Pharmatherm pallet shipping systems are designed to offer optimal thermal protection for pharmaceutical products.
  • Large internal volume
  • Optimal thermal protection
  • Pallet sized shipper
These shipping systems are pre-qualified to maintain payload integrity during shipments for up to 96 hours. The range is tested against Intelsius® demanding high performance dry ice temperature and high performance 2 to 8 °C profiles.