Creatinine

Supplier: DIAGNOSTIC SYSTEMS DIASYS

Synonyms: 2-Amino-1-methyl-1,5-dihydro-4H-imidazol-4-one

117119910021
DIAS117119910021EA 0 EUR
DIAS117119910021
Creatinine
Creatinine
Formula: C₄H₇N₃O
MW: 113.12 g/mol
MDL Number: MFCD00059730
CAS Number: 60-27-5
EINECS: 200-466-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR