Teaching camera
Supplier: DIDALAB
POF010810
DIDAPOF010810EA 1140 EUR
DIDAPOF010810
Teaching camera
Cameras
The pedagogical model of the digital camera can be put on a Ø-10 mm rod or on a camera’s stand.
  • Pentax lenses: One with a fixed 50-mm focus, the other with a 28 to 70-mm focus
  • Focus, aperture, time exposure, field depth, magnification, sensor, sensibility and accuracy
  • Image reflection with mirror to understand the Reflex aiming
By setting the lens, the students can understand the field depth and the digital aperture principle.
Delivery information: Supplied with specific software and a complete manual.