Disinfectants, ClearKlens Cleansinald, sterile
Supplier: Diversey
100848254 100848253
DIVY100848254EA 328 EUR
DIVY100848254 DIVY100848253
Disinfectants, ClearKlens Cleansinald, sterile
Cleaning Agents Disinfectants
Surface active disinfectant based on a blend of quaternary ammonium and alkyl amines.
  • 0,2 µm filtered, aseptically filled and double bagged
  • Broad spectrum of activity
  • Supplied with certificate of analysis and sterility where applicable
  • Compatible with ClearKlens Tego® 2000, ClearKlens Tego® 2001 and ClearKlens Bi-Spore as a rotational process
Certifikace: Passes CEN 1276, CEN 1650 and CEN 13697
Upozornení: Use biocides safely. Always read the label and product information before use.
Articles may not be available in all countries. Please contactMore Product Information