Hand disinfectant, Soft Care Des E spray H5, W3646
Supplier: Diversey
100860440
DIVY100860440EA 162 EUR
DIVY100860440
Hand disinfectant, Soft Care Des E spray H5, W3646
Skincare Skin Disinfectants Liquid and Foam Skin Disinfectants
Soft Care Des E spray is a hand disinfectant based on alcohol (71,58%) and is applied without water. Soft Care Des E spray is suitable for use in all areas, including healthcare and the food sector.
  • Fragrance-free, eliminating the risk of food contamination and making it suitable for use in the food industry
  • Low viscosity, making it suitable for use in a wide variety of hygiene equipment
  • Suitable for surgical disinfection
  • Contains an effective moisturiser that prevents skin irritation and dryness
  • Liquid suitable for spray application