Safety cabinet type 90 UTS ergo line S with classical door technology. For the storage of flammable liquids. Type-tested by MPA/ TÜV SÜD according to DIN EN 14470-1, laboratory furniture standard DIN EN 14727, EK5/AK4 09-10:2009 and TRGS 510 Annex 3 1 * 1 items
Supplier: DUPERTHAL
Safety cabinet type 90 UTS ergo line S with classical door technology. For the storage of flammable liquids. Type-tested by MPA/ TÜV SÜD according to DIN EN 14470-1, laboratory furniture standard DIN EN 14727, EK5/AK4 09-10:2009 and TRGS 510 Annex 3 1 * 1 items