Safety underbench cabinets, type 90, UTS ergo line LD-5, 500 mm depth
Supplier: DUPERTHAL
29-061159-405 29-061159-005 29-061157-505 29-061157-405 29-061157-005
DUEH29-061159-405EA 2493.33 EUR
DUEH29-061159-405 DUEH29-061159-005 DUEH29-061157-505 DUEH29-061157-405 DUEH29-061157-005
Safety underbench cabinets, type 90, UTS ergo line LD-5, 500 mm depth
Furniture Storage Cabinets
These safety cabinets are fire resistant up to 90 minutes. These under bench cabinets are equipped with wing doors and/or drawers for the storage of hazardous substances in working spaces.
  • Easy moving, single-handed operation of the doors and drawers
  • All UTS ergo line models are fitted with a cylinder lock as a standard
  • Two door technologies are available: the classical on or the push-to open wing door technology (the easy moving safety doors are opened by applying a gentle push to the front)
Drawers or wing doors remain open in any position andMore Product Information