Gas cylinder depots, for storing gas cylinders outdoor
Supplier: DUPERTHAL
70-201340-001 70-201340-003 70-201040-001 70-200740-003 70-201040-003 70-200740-001
DUEH70-201340-001EA 1479.66 EUR
DUEH70-201340-001 DUEH70-201340-003 DUEH70-201040-001 DUEH70-200740-003 DUEH70-201040-003 DUEH70-200740-001
Gas cylinder depots, for storing gas cylinders outdoor
Furniture Storage Cabinets
Gas cylinder depots are used for storing gas cylinders outdoors in accordance with the regulations.
  • Robust construction made of sheet steel, material thickness 1,5 mm
  • Three-point locking system with door handle with security cylinder lock at an ergonomically favourable height
  • Two horizontal installation rails for mounting fittings
  • Prepared for ground fixing and earthing possibility on the ceiling
  • Natural ventilation and labelling with warning symbol W19
These gas cylinder depots have good corrosion resistance with polyester powder coating RAL 9002More Product Information