Manganese(II) chloride tetrahydrate 98-101 ACS

Supplier: VWR Chemicals
Warning

Synonyms: Manganese dichloride tetrahydrate, Manganese chloride (tetrahydrate), Manganese(II) chloride tetrahydrate, Manganous chloride tetrahydrate, Manganese chloride tetrahydrate, Manganese Chloride, Tetrahydrate

E107-100GEA 113 EUR
E107-100G E107-500G
Manganese(II) chloride tetrahydrate 98-101 ACS
Manganese (II) chloride tetrahydrate
Formula: MnCl₂·4H₂O
MW: 197.9 g/mol
Boiling Pt: 1190
Melting Pt: 58…59 °C
Density: 2.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149792
CAS Number: 13446-34-9
EINECS: 231-869-6
UN: 3077
ADR: 9 III
Merck Index: 13,05751

Order Now

Specification Test Results

Calcium <0.005%
Heavy Metals ≤0.0005%
Insolubles ≤0.005%
Iron ≤0.0005%
Magnesium ≤0.005%
pH (5%, Water) @25°C 3.5 - 6
Potassium ≤0.01%
Purity 98 - 101%
Sodium ≤0.05%
Sulphate ≤0.005%
Zinc ≤0.005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR