Winlog.Validation Evaluation Software for Ebro Data Loggers EBI 12, EBI 11 and EBI 16
Supplier: EBRO
1340-2470
EBRO1340-2470EA 5240 EUR
EBRO1340-2470
Winlog.Validation Evaluation Software for Ebro Data Loggers EBI 12, EBI 11 and EBI 16
Software Laboratory Software
The easy-to-use software products are suitable for programming and read-out of Ebro® data loggers as well as for the evaluation of the measured values. For routine checks and validation.
  • User-friendly
  • Highly accurate measurements
  • Automatic report generation
  • Automatic user-defined calculations
  • Automatic recognition of process cycles
  • Current version: 4.0
Save time and effort for the regularly required process validation with the help of the powerful reporting and evaluation software Winlog.validation. The automatic recognition of the individual processMore Product Information