1,1,1-Trichloro-2-methyl-2-propanol hemihydrate

Supplier: EHRENSTORFER

Synonyms: 1,1,1-Trichloro-2-methyl-2-propanol Hemihydrate, Chlorbutanol hemihydrate, Chlorobutanol hemihydrate, Chlorobutanol hydrous

C10020000
EHERC10020000EA 48.4 EUR
EHERC10020000
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate
Formula: C₈H₁₆Cl₆O₃
MW: 372.93 g/mol
Boiling Pt: 167 °C (1013 hPa)
Melting Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD02179352
CAS Number: 6001-64-5
EINECS: 200-317-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR