Bithionol

Supplier: EHRENSTORFER

Synonyms: Bithionol, Bis(3,5-dichloro-2-hydroxyphenyl) sulphide, 2,2'-Thiobis(4,6-dichlorophenol), 6,6'-Thiobis(2,4-dichlorophenol)

C10660500
EHERC10660500EA 72 EUR
EHERC10660500
Bithionol
Bithionol
Formula: C₁₂H₆Cl₄O₂S
MW: 356.0518 g/mol
MDL Number: MFCD00055727
CAS Number: 97-18-7
EINECS: 202-565-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR