6-Chloro-o-toluidine

Supplier: EHRENSTORFER
Warning

Synonyms: 2-Chloro-6-methylaniline, 2-Amino-3-chlorotoluene

C11429600
EHERC11429600EA 47.2 EUR
EHERC11429600
6-Chloro-o-toluidine
6-Chloro-o-toluidine
Formula: C₇H₈ClN
MW: 141.6 g/mol
Boiling Pt: 215
Melting Pt: 2
Density: 1.152 g/cm³ (20 °C)
MDL Number: MFCD00007679
CAS Number: 87-63-8
EINECS: 201-759-2
UN: 3429
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR