Dipropyleneglycol monomethyl ether (mixture of isomers)

Supplier: EHRENSTORFER

Synonyms: Dipropylene glycol monomethyl ether, 1-((1-Methoxypropan-2-yl)oxy)propan-2-ol

C12938500
EHERC12938500EA 36.6 EUR
EHERC12938500
Dipropyleneglycol monomethyl ether (mixture of isomers)
Dipropyleneglycol monomethyl ether (mixture of isomers)
Formula: C₇H₁₆O₃
MW: 148.2001 g/mol
MDL Number: MFCD00059604
CAS Number: 34590-94-8
EINECS: 252-104-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR