2,3,4,5-Tetrachloroaniline

Supplier: EHRENSTORFER

C17330400
EHERC17330400EA 741 EUR
EHERC17330400
2,3,4,5-Tetrachloroaniline
2,3,4,5-Tetrachloroaniline
Formula: C₆H₃Cl₄N
MW: 230.91 g/mol
CAS Number: 634-83-3
EINECS: 211-216-1
UN: 1648
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR