1,2,3,4-Tetrachlorobenzene

Supplier: EHRENSTORFER
Warning

C17353400
EHERC17353400EA 43.4 EUR
EHERC17353400
1,2,3,4-Tetrachlorobenzene
1,2,3,4-Tetrachlorobenzene
Formula: C₆H₂Cl₄
MW: 215.89 g/mol
Boiling Pt: 254 °C (1013 hPa)
Melting Pt: 42…45 °C
Density: 1,73 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000538
CAS Number: 634-66-2
EINECS: 211-214-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR