α,2,6-Trichlorotoluene

Supplier: EHRENSTORFER
Danger

Synonyms: 1-(Chloromethyl)-2,6-dichlorobenzene, 2,6-Dichlorobenzyl Chloride, 2,6-Dichlorobenzyl chloride, 1,3-Dichloro-2-(chloromethyl)benzene

C17787600
EHERC17787600EA 36.9 EUR
EHERC17787600
α,2,6-Trichlorotoluene
α,2,6-Trichlorotoluene
Formula: C₇H₅Cl₃
MW: 195.47 g/mol
Boiling Pt: 248 °C (1013 hPa)
Melting Pt: 49…50 °C
MDL Number: MFCD00000897
CAS Number: 2014-83-7
EINECS: 217-940-4
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR