n-Undecane

Supplier: EHRENSTORFER
Danger

Synonyms: Undecane

C17896300
EHERC17896300EA 70.7 EUR
EHERC17896300
n-Undecane
n-Undecane
Formula: H₃C(CH₂)₉CH₃
MW: 156.31 g/mol
Boiling Pt: 196
Melting Pt: –26.7 °C
Density: 0.7416 g/cm³ (20 °C)
Flash Pt: 60
Storage Temperature: Ambient
MDL Number: MFCD00008959
CAS Number: 1120-21-4
EINECS: 214-300-6
UN: 2330
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR