4,4'-Dibromobiphenyl (PBB No. 15)

Supplier: EHRENSTORFER
Warning

Synonyms: PBB No. 15

C21001500
EHERC21001500EA 47.2 EUR
EHERC21001500
4,4'-Dibromobiphenyl (PBB No. 15)
4,4'-Dibromobiphenyl (PBB No. 15)
Formula: C₁₂H₈Br₂
MW: 312 g/mol
Boiling Pt: 357 °C (1013 hPa)
Melting Pt: 164 °C
Density: 1.867 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000101
CAS Number: 92-86-4
EINECS: 202-198-6
UN: 3152
ADR: 9,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR