Single wafer tray, H 20 series, 3" (76,2 mm)
Supplier: ENTEGRIS
H202000011415 H202000021201
ENTEH202000011415EA 0 EUR
ENTEH202000011415 ENTEH202000021201
Single wafer tray, H 20 series, 3" (76,2 mm)
Wafer Handling Supplies Wafer Trays
Wafer handling trays to safely transport and protect silicon wafers.
  • For internal handling of single wafers
  • Conical pocket holds one wafer facedown contacting only the wafer’s edges
  • Tweezer slots on tray bottom ease wafer removal
  • Stackable for efficient use of storage space, either tray on tray or cover on tray