5-Methylcytosine DNA ELISA Kit
Supplier: ENZO LIFE SCIENCES
ADI900224A0001
ENZOADI900224A0001EA 701 EUR
ENZOADI900224A0001
5-Methylcytosine DNA ELISA Kit
Assays ELISAs
The 5-Methylcytosine DNA ELISA kit is a complete, colorimetric, indirect immunoassay kit for the quantitative determination of 5-methylcytosine in DNA with results in less than three hours.
  • Rapid – fully quantitative results in <3 hours
  • Convenient – user-friendly alternative to mass spectrometry
  • Simple – efficient and well-established protocol
  • High-throughput – 96 well format ideal for global 5-methylcytosine detection in DNA
The ability to efficiently detect and quantify DNA methylation (i.e., 5-methylcytosine) has become essential for epigenetic-based research. To date, a number of methods have been developed for this purpose including high-performance capillary electrophoresis, bisulfite sequencing, and methylated DNA immunoprecipitation.
Shipping conditions: Dry ice