11-dehydro-TXB2 ELISA Kits

Supplier: ENZO LIFE SCIENCES

ADI901092
ENZOADI901092EA 2110 EUR
ENZOADI901092
11-dehydro-TXB2 ELISA Kits
Assays ELISAs

For the quantitative determination of 11-dehydro-TXB2 in culture supernatants and urine from any species. Cited samples include serum.


The 11-dehydro-TXB2 EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Absorbance is read at 405 nm. This kit offers ultrasensitive quantification of 11-d-TXB2 in a variety of sample matrices with negligible reactivity with related molecules in a convenient 96-well format offers higher throughput analysis compared to conventional analytical methods.


Informace pro objednávání: Contains GxR Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-140 (1x96 wells).

New Product

Order Now

SPECIFICATIONS

Environmentally Preferable
Target protein 11-dehydro-TXB2
Primary antibody reactivity
Assay Type Competitive
Host
Sensitivity 4,31 pg/ml (range 9,8 - 10,000 pg/ml)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR