Anti-PDI Rabbit Polyclonal Antibody

Supplier: ENZO LIFE SCIENCES
ADISPA890D
ENZOADISPA890DEA 241 EUR
ENZOADISPA890D
Anti-PDI Rabbit Polyclonal Antibody
Antibodies
The mammalian PDI (Protein disulfide-isomerase) family encompasses several highly divergent proteins which are involved in the processing and maturation of secretory proteins in the endoplasmic reticulum by catalyzing the rearrangement of disulfide bonds.

  • For Immunocytochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, rat, bovine, dog, guinea pig, hamster, monkey, pig, sheep and Xenopus PDI. Detects a band of ~58kDa by Western blot.

Type: Primary
Antigen: PDI
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR