L(-)-Glutathione (reduced form)

Supplier: ENZO LIFE SCIENCES

Synonyms: L-Glutathione Reduced, GSH, (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, Glutathione Reduced

ALX157002G005 ALX157002G025
ENZOALX157002G005EA 55 EUR
ENZOALX157002G005 ENZOALX157002G025
L(-)-Glutathione (reduced form)
L(-)-Glutathione (reduced form)
Formula: C₁₀H₁₇N₃O₆S
MW: 307.33 g/mol
Melting Pt: 185…195 °C
Density: 1,475 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00065939
CAS Number: 70-18-8
EINECS: 200-725-4
Merck Index: 13,04488

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR