Human;Rat recombinant COMP:Angiopoietin-1 (from CHO cells)

Supplier: ENZO LIFE SCIENCES

ALX201314C010 ALX201314C050 ALX201314C500
ENZOALX201314C010EA 752 EUR
ENZOALX201314C010 ENZOALX201314C050 ENZOALX201314C500
Human;Rat recombinant COMP:Angiopoietin-1 (from CHO cells)
Proteins and Peptides

Soluble, stable and more potent Ang-1 variant.


Produced in CHO cells. The fibrinogen-like domain (aa 284-498) of human Ang-1 (angiopoietin-1) is fused at the N-terminus to the coiled-coil domain of rat COMP (cartilage oligomeric matrix protein) and to a FLAG®-tag.


Soluble, stable and more potent Ang-1 variant.

New Product

Order Now

SPECIFICATIONS

Protein/peptide name COMP:Angiopoietin-1 COMP:Angiopoietin-1
Environmentally Preferable
Protein/peptide type Recombinant Recombinant
Species Human,Rat Human,Rat
Source CHO cells CHO cells
Biological activity Induces phosphorylation of Tie-2 in primary cultured human and mouse vascular endothelial cells. More potent than native Ang-1.
Purity >90% (SDS-PAGE)
Concentration ~0,3 mg/ml
Formulation Liquid. 0,2 µM-filtered solution in PBS containing 0,01% CHAPS.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR