(-)-Eburnamonine ≥99%

Supplier: ENZO LIFE SCIENCES

Synonyms: (41S,13aS)-13a-Ethyl-2,3,5,6,13,13a-hexahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridin-12(41H)-one, (3α,16α)-Eburnamenin-14(15H)-one, vincamone, vinburnine

ALX350216M100
ENZOALX350216M100EA 160 EUR
ENZOALX350216M100
(-)-Eburnamonine ≥99%
(-)-Eburnamonine

Antihypoxic. Vasodilator.


Cerebral metabolic stimulant; shows antihypoxic effects.


Soluble in chloroform.

Formula: C₁₉H₂₂N₂O
MW: 294.4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00064309
CAS Number: 4880-88-0
EINECS: 225-490-5
Merck Index: 14,03487
New Product

Order Now

Specification Test Results

Purity ≥99% (Assay)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR