Ciprofloxacin 98

Supplier: ENZO LIFE SCIENCES

Synonyms: 1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid, 1-Cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic Acid

ALX380287G005 ALX380287G025
ENZOALX380287G005EA 50.6 EUR
ENZOALX380287G005 ENZOALX380287G025
Ciprofloxacin 98
Ciprofloxacin

Fluoroquinolone antibacterial agent. Inhibits bacterial DNA gyrase (topoisomerase). Inhibits cell division and causes double-strand breaks in the bacterial chromosome. Broad-spectrum antibiotic effective against Gram-positive, Gram-negative and anaerobic bacteria.


  • Effective against Mycobacterium tuberculosis
  • Potentially anxiogenic


White to off-white powder


Soluble in dilute hydrochloric acid; insoluble in water


Balení: Shipping temperature: Ambient

Formula: C₁₇H₁₈FN₃O₃
MW: 331.35 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00185755
CAS Number: 85721-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR