WS-12 ≥98% (by HPLC)

Supplier: ENZO LIFE SCIENCES
Danger

Synonyms: AVX-012, (1R,2S,5R)-2-Isopropyl-N-(4-methoxyphenyl)-5-methylcyclohexane-1-carboxamide

ALX420041M025
ENZOALX420041M025EA 464 EUR
ENZOALX420041M025
WS-12 ≥98% (by HPLC)
WS-12

TRMP8 agonist.


Cooling agent. High-affinity TRPM8 (cold menthol receptor 1; CMR1) agonist. Has an EC₅₀ value ~2000 times lower than that of (-)-menthol.


Soluble in 100% ethanol, methanol, ethyl acetate, dimethyl formamide or acetonitrile; insoluble in water or DMSO.

Formula: C₁₈H₂₇NO₂
MW: 289.4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD11100191
CAS Number: 68489-09-8
New Product

Order Now

Specification Test Results

Purity ≥98% (HPLC)
Identity Determined by ¹H-NMR and MS

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR