Sodium chloride 0.9% physiological saline endotoxin-free, sterile

Supplier: ENZO LIFE SCIENCES

Synonyms: Saline, NaCl, Sea salt, Halite

ALX505009LD15
ENZOALX505009LD15EA 59.4 EUR
ENZOALX505009LD15
Sodium chloride 0.9% physiological saline endotoxin-free, sterile
Sodium chloride

Sterile and endotoxin-free physiological saline for use with TLRgrade™ or endotoxin-free grade reagents.


Product has been subjected to multiple rounds of LPS removal by adsorption with activated charcoal.

Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
Storage Temperature: Refrigerator
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Merck Index: 14,08599

Order Now

Specification Test Results

Endotoxin content <0.0002 EU/µg (pyrotell gel-clot endotoxin test)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR