Pilocarpine hydrochloride

Supplier: ENZO LIFE SCIENCES

Synonyms: (+)-Pilocarpine hydrochloride

ALX550092G001
ENZOALX550092G001EA 66 EUR
ENZOALX550092G001
Pilocarpine hydrochloride
Pilocarpine hydrochloride
Formula: C₁₁H₁₇ClN₂O₂
MDL Number: MFCD00012722
CAS Number: 54-71-7
EINECS: 200-212-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR