MPLA 1mg/ml (from Salmonella minnesota) ≥99.9%, TLRgrade™

Supplier: ENZO LIFE SCIENCES

Synonyms: Monophosphoryl Lipid A

ALX581202L001 ALX581202R500
ENZOALX581202L001EA 601 EUR
ENZOALX581202L001 ENZOALX581202R500
MPLA 1mg/ml (from Salmonella minnesota) ≥99.9%, TLRgrade™
MPLA

TLR4 activator.


Isolated from Salmonella minnesota R595 (Re mutant). Prepared by acid hydrolysis from purified Salmonella minnesota R595 (Re) LPS (isolated by a modification of the PCP extraction method), converted to the uniform salt form.


Activates Toll-like receptor (TLR) 4. Does not activate TLR2 as determined with splenocytes and macrophages from TLR4 deficient mice. Exhibits reduced endotoxic and pyrogenic activity compared to lipid A and LPS.

Order Now

Specification Test Results

Concentration 1 mg/ml
Formulation Liquid. Sterile, ready-to-use solution in pyrogen-free double distilled water
Purity ≥99.9% (regarding protein or DNA contaminants, KDO not detectable)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR