Lucigenin 97 HPLC_ASSAY_METHOD

Supplier: ENZO LIFE SCIENCES

Synonyms: Bis(N-methylacridinium) nitrate, N,N′-Dimethyl-9,9′-biacridinium dinitrate, 10,10'-Dimethyl-9,9'-biacridinium dinitrate

ALX620061M050
ENZOALX620061M050EA 53.9 EUR
ENZOALX620061M050
Lucigenin 97 HPLC_ASSAY_METHOD
Lucigenin

ROS dye


Chemiluminescent probe for the detection of superoxide in biological systems.

Formula: C₂₈H₂₂N₄O₆
MW: 510.51 g/mol
MDL Number: MFCD00011925
CAS Number: 2315-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR