Podophyllotoxin ≥98% (by HPLC)

Supplier: ENZO LIFE SCIENCES

Synonyms: (10R,11R,15R,16R)-16-hydroxy-10-(3,4,5-trimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.03,7.011,15]hexadeca-1,3(7),8-trien-12-one, Condylox, PPT, (-)-Podophyllotoxin, Podofilox, Condyline, NSC 24818

ALX630086M100
ENZOALX630086M100EA 57.2 EUR
ENZOALX630086M100
Podophyllotoxin ≥98% (by HPLC)
Podophyllotoxin

Topoisomerase inhibitor


Potent inhibitor of microtubule assembly. Antineoplastic glucoside. Antitumor agent. DNA topoisomerase II inhibitor.


Isolated from Podophyllum peltatum.

Formula: C₂₂H₂₂O₈
MW: 414.41 g/mol
MDL Number: MFCD00075290
CAS Number: 518-28-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR