Moniliformin sodium salt 98 TLC_ASSAY_METHOD

Supplier: ENZO LIFE SCIENCES
Danger

ALX630111M001
ENZOALX630111M001EA 352 EUR
ENZOALX630111M001
Moniliformin sodium salt 98 TLC_ASSAY_METHOD
Moniliformin sodium salt

Mycotoxin.


Soluble in water, methanol or DMSO.

Formula: C₄HO₃·Na
MW: 120.1 g/mol
Storage Temperature: Freezer
CAS Number: 71376-34-6

Order Now

Specification Test Results

Source Isolated from Fusarium moniliforme
Purity ≥98% (TLC)
Identity Identity determined by NMR


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR