Anti-IL1R2 Mouse Monoclonal Antibody

Supplier: ENZO LIFE SCIENCES
ALX804461C100
ENZOALX804461C100EA 598 EUR
ENZOALX804461C100
Anti-IL1R2 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For ELISA, Flow Cytometry, Immunohistochemistry, Western Blot

Recognizes human IL-1RII.

Type: Primary
Antigen: IL1R2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol IL1R2
Environmentally Preferable
Antigen name Interleukin-1 receptor 2
Antigen synonyms IL-2RB|CD122|p75|IL-2R subunit beta|high affinity IL-2 receptor beta subunit|beta|P70-75|IL15RB|IL-2 receptor subunit beta|interleukin-2 receptor subunit beta|interleukin 15 receptor|high affinity IL-2 receptor subunit beta
Antibody type Primary
Clonality Monoclonal
Reactivity
Host Mouse
Isotype IgG1
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Flow cytometry Yes
Form Liquid. Purified antibody in PBS containing 0,1% sodium azide
Concentration 1 mg/ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR