Anti-IDO1 Rat Monoclonal Antibody [clone: mIDO-48]
Supplier: ENZO LIFE SCIENCES
ALX8049010100
ENZOALX8049010100EA 305 EUR
ENZOALX8049010100
Anti-IDO1 Rat Monoclonal Antibody [clone: mIDO-48]
Antibodies
Indoleamine 2,3-dioxygenase (IDO) is an IFN-γ inducible gene. It catalyzes the degradation of the essential amino acid L-tryptophan to N-formylkynurenine. IDO has been implicated in protection against intracellular and extracellular pathogens. The mIDO-48 monoclonal antibody recognizes mouse IDO also known as Indolamine 2,3-dioxygenase, Indole 2,3-dioxygenase, and Indoleamine-pyrrole 2,3-dioxygenase. This antibody does not appear to react with human IDO. IDO is a ubiquitously expressed cytoplasm protein with a predicted molecular weight approximately 45 kDa. Indoleamine 2,3-dioxygenase (IDO)More Product Information