Meclofenamic acid sodium salt 98 HPLC_ASSAY_METHOD

Supplier: ENZO LIFE SCIENCES

Synonyms: Sodium 2-((2,6-dichloro-3-methylphenyl)amino)benzoate

BMLEI1630005
ENZOBMLEI1630005EA 93.5 EUR
ENZOBMLEI1630005
Meclofenamic acid sodium salt 98 HPLC_ASSAY_METHOD
Meclofenamic acid sodium salt

Dual 5-lipoxygenase (IC50=46.8 µM) and cyclooxygenase I and II inhibitor (IC50=0.6 µM). Clinically useful NSAID.


  • COX inhibitor


White solid.


Soluble in water (15mg/ml slightly cloudy).

Formula: C₁₄H₁₀Cl₂NNaO₂
MW: 318.1 g/mol
Melting Pt: 289…291 °C
Storage Temperature: Ambient
MDL Number: MFCD00077376
CAS Number: 6385-02-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ded15ca6dd8918e259a8b51f64d2a35bb74e0ba&vid=f2e622785217d3b8396a817e0d581712da06142d&prid=web&clid=SAPDAM
New Product

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR