Ozagrel ≥98%

Supplier: ENZO LIFE SCIENCES

Synonyms: (E)-3-(4-((1H-Imidazol-1-yl)methyl)phenyl)acrylic acid

BMLEI3780010
ENZOBMLEI3780010EA 86.9 EUR
ENZOBMLEI3780010
Ozagrel ≥98%
Ozagrel

Potent and selective thromboxane A2 synthase inhibitor (IC50=11nM).


  • Thromboxane synthase inhibitor


Potent and selective thromboxane A2 synthase inhibitor (IC50=11nM).

Formula: C₁₃H₁₂N₂O₂
MW: 228.2466 g/mol
MDL Number: MFCD00868231
CAS Number: 82571-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR