PF-622 ≥98% (by TLC)

Supplier: ENZO LIFE SCIENCES

BMLEI4020005
ENZOBMLEI4020005EA 127 EUR
ENZOBMLEI4020005
PF-622 ≥98% (by TLC)
PF-622

FAAH inhibitor


Novel, potent and selective inhibitor of fatty acid amide hydrolase (FAAH). Inhibits FAAH in a time-dependent manner by covalently modifying the enzyme's active site serine (IC50=33nM). It is completely selective for FAAH relative to other mammalian serine hydrolases.

Formula: C₂₁H₂₂N₄O
MW: 346.4 g/mol
CAS Number: 898235-65-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1446c155a50b62c52a369ee03526d85599e5e396&vid=86098ad3277348bba51be571d71325bf63671433&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR