MeOSuc-AAPV-AMC
Supplier: ENZO LIFE SCIENCES
BMLP2240005
ENZOBMLP2240005EA 165 EUR
ENZOBMLP2240005
MeOSuc-AAPV-AMC
Proteins and Peptides
Elastase substrate
Highly sensitive substrate for neutrophil elastase (leukocyte elastase) (Km=130 µM), but not granzyme H. This substrate can be used for in vitro assays with purified enzyme, on live cells, or in conditioned medium. Ex: 380 nm, Em: 460 nm.