TACE substrate II
Supplier: ENZO LIFE SCIENCES
BMLP2280100
ENZOBMLP2280100EA 161 EUR
ENZOBMLP2280100
TACE substrate II
Proteins and Peptides
Sensitive fluorogenic substrate for TACE (TNFalpha converting enzyme, ADAM17). Also cleaved by ADAM10 and likely other ADAMs and MMPs. The kcat/Km of TACE for this substrate is 7-fold higher than for the same peptide with the Mca-Dpa fluorophore-quencher pair. FAM fluorescence is quenched by the TAMRA group until cleavage separates them. Ex.: 485 nm, Em.: 535 nm, although the following Ex/Em can also be used: 490,494/515,520 nm. This substrate is useful for inhibitor screening, kinetic analysis, and cellular activity assay.